Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NAV2 EBGEX Assays ›
Gene Symbol:
NAV2
Gene Name:
neuron navigator 2
Gene Aliases:
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
Chromosome Location:
Chr.11: 19345236 - 20121601 on Build GRCh38
UniGene ID:
Hs.502116
Species:
Human
Species Specific ID (Flybase ID):
-
Type:
1
Detection Label:
Alexa Fluor 546
ProductType:
ViewRNA Cell Probe Set
Assay ID
VA1-3012896-VC
Size
M: 440 uL
L: 1.44 mL
Availability
Made To Order
Catalog #
VX-01
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: NAV2
Interrogated Sequence
Translated Protein
IMAGE Clone ID
RefSeq
NM_001111018.2
NP_001104488.1
NM_001111019.3
NP_001104489.1
NM_001244963.2
NP_001231892.1
NM_145117.5
NP_660093.2
NM_182964.6
NP_892009.3
XM_005253214.5
XP_005253271.1
XM_006718364.5
XP_006718427.1
XM_006718365.5
XP_006718428.1
XM_006718366.5
XP_006718429.1
XM_011520444.4
XP_011518746.1
XM_011520445.4
XP_011518747.1
XM_011520446.4
XP_011518748.1
XM_011520447.4
XP_011518749.1
XM_011520448.4
XP_011518750.1
XM_011520449.4
XP_011518751.1
XM_011520450.4
XP_011518752.1
XM_017018520.3
XP_016874009.1
XM_017018521.3
XP_016874010.1
XM_017018522.2
XP_016874011.1
XM_017018524.3
XP_016874013.1
XM_024448758.2
XP_024304526.1
XM_047427809.1
XP_047283765.1
XM_047427810.1
XP_047283766.1
XM_047427811.1
XP_047283767.1
XM_047427812.1
XP_047283768.1
XM_047427813.1
XP_047283769.1
XM_047427814.1
XP_047283770.1
XM_047427815.1
XP_047283771.1
XM_047427816.1
XP_047283772.1
XM_047427817.1
XP_047283773.1
XM_047427818.1
XP_047283774.1
XM_047427819.1
XP_047283775.1
XM_047427820.1
XP_047283776.1
XM_047427821.1
XP_047283777.1
XM_047427822.1
XP_047283778.1
XM_047427823.1
XP_047283779.1
XM_047427824.1
XP_047283780.1
XM_047427825.1
XP_047283781.1
XM_047427826.1
XP_047283782.1
XM_047427827.1
XP_047283783.1
XM_047427828.1
XP_047283784.1
XM_047427829.1
XP_047283785.1
XM_047427830.1
XP_047283786.1
XM_047427831.1
XP_047283787.1
XM_047427833.1
XP_047283789.1
XM_047427834.1
XP_047283790.1
XM_047427835.1
XP_047283791.1
XM_047427836.1
XP_047283792.1
XM_047427837.1
XP_047283793.1
XM_047427838.1
XP_047283794.1
GenBank mRNA
AA451836.1
-
AB037840.3
-
AB063115.1
-
AB063116.1
-
AF466143.1
-
AF466144.1
-
AJ488102.1
-
AJ488202.1
-
AJ488203.1
-
AJ488204.1
-
AJ488205.1
-
AJ488206.1
-
AJ488207.1
-
AJ488208.1
-
AK001892.1
-
AK074287.1
-
AK096037.1
-
AK290458.1
-
AK298346.1
-
AK307574.1
-
BC016054.1
-
BC107690.1
-
BC150290.1
-
BE818973.1
-
BI831180.1
-
BX459843.2
-
BX485661.1
-
R23148.1
-
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of systemic arterial blood pressure by baroreceptor feedback
nervous system development
sensory perception of sound
sensory perception of smell
locomotory behavior
optic nerve development
glossopharyngeal nerve development
vagus nerve development
cerebellar cortex development
neurogenesis
nucleotide binding
helicase activity
protein binding
ATP binding
heparin binding
hydrolase activity
ATPase activity
3'-5' DNA helicase activity
Back To Top
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline