Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDK11A EBGEX Assays ›
          Gene Symbol:
          CDK11A
          Gene Name:
          cyclin dependent kinase 11A
          Gene Aliases:
          CDC2L2, CDC2L3, CDK11-p110, CDK11-p46, CDK11-p58, PITSLRE, p58GTA
          Chromosome Location:
          Chr.1: 1702383 - 1724565 on Build GRCh38
          UniGene ID:
          Hs.651228
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Type:
          1
          Detection Label:
          FastRed
          ProductType:
          ViewRNA Tissue Probe Set
          Assay ID VA1-3020420-VT
          Size
          Availability Made To Order
          Catalog # VX-01
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CDK11A
          Interrogated Sequence Translated Protein IMAGE Clone ID
          RefSeq NM_001313896.1 NP_001300825.1
          NM_001313982.1 NP_001300911.1
          NM_024011.3 NP_076916.2
          NM_033529.3 NP_277071.2
          NR_132739.1
          NR_132740.1
          GenBank mRNA AF067518.1 -
          AF067519.1 -
          AF067520.1 -
          AF067521.1 -
          AF067522.1 -
          AF067523.1 -
          AF067524.1 -
          AF067526.1 -
          AF067527.1 -
          AF067528.1 -
          AF067529.1 -
          AK301495.1 -
          AK303446.1 -
          AK303451.1 -
          AL043340.1 -
          BC014464.1 -
          BC028187.1 -
          BC073855.1 -
          BC094827.1 -
          BC110905.1 - 6151813
          DC388505.1 -
          ENST00000407249.0 -
          U04819.1 -
          U07704.1 -
          U07705.1 -

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          regulation of transcription, DNA-templated
          protein phosphorylation
          apoptotic process
          mitotic nuclear division
          regulation of mRNA processing
          regulation of cell cycle
          protein kinase activity
          protein serine/threonine kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          ATP binding

          Back To Top
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2lcrw:80/100.66.78.62:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0