Assay Details
Gene Symbol: PIK3CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001256045.1 | NP_001242974.1 | ||||
NM_006219.2 | NP_006210.1 | |||||
XM_005247530.2 | XP_005247587.1 | |||||
XM_006713659.3 | XP_006713722.1 | |||||
XM_011512895.2 | XP_011511197.1 | |||||
XM_011512896.1 | XP_011511198.1 | |||||
XM_017006619.1 | XP_016862108.1 | |||||
XM_017006620.1 | XP_016862109.1 | |||||
XM_017006621.1 | XP_016862110.1 | |||||
GenBank mRNA | AI623206.1 | - | ||||
AK293754.1 | - | |||||
AK302937.1 | - | |||||
BC003393.1 | - | |||||
BC022049.2 | - | |||||
BC033948.2 | - | |||||
BC035432.2 | - | |||||
BC045790.1 | - | |||||
BC114432.1 | - | 40008544 | ||||
CR749357.1 | - | |||||
ENST00000477593.0 | - | |||||
S67334.1 | - |
