Search
Search

THPO| Gene Symbol: THPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_000460.3 | NP_000451.1 | ||||
| NM_001177597.2 | NP_001171068.1 | |||||
| NM_001177598.2 | NP_001171069.1 | |||||
| NM_001289997.1 | NP_001276926.1 | |||||
| NM_001289998.1 | NP_001276927.1 | |||||
| NM_001290003.1 | NP_001276932.1 | |||||
| NM_001290022.1 | NP_001276951.1 | |||||
| NM_001290026.1 | NP_001276955.1 | |||||
| NM_001290027.1 | NP_001276956.1 | |||||
| NM_001290028.1 | NP_001276957.1 | |||||
| XM_017007107.1 | XP_016862596.1 | |||||
| GenBank mRNA | AA479058.1 | - | ||||
| AA480029.1 | - | |||||
| AB014681.1 | - | |||||
| AB014682.1 | - | |||||
| AB014683.1 | - | |||||
| AB102890.1 | - | |||||
| AB102892.1 | - | |||||
| AB102894.1 | - | |||||
| AK311002.1 | - | |||||
| BC130322.1 | - | 40146353 | ||||
| BC136338.1 | - | 9020325 | ||||
| BC143982.1 | - | 9052500 | ||||
| BX108191.1 | - | |||||
| D32047.1 | - | |||||
| ENST00000204615.0 | - | |||||
| L33410.1 | - | |||||
| L36052.1 | - | |||||
| U11025.1 | - | |||||
| U42977.1 | - | |||||
| U59493.1 | - | |||||
| U59494.1 | - | |||||
| U59495.1 | - | |||||