Search
Search

EVA1C| Gene Symbol: EVA1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001286556.2 | NP_001273485.1 | ||||
| NM_001320744.2 | NP_001307673.1 | |||||
| NM_001320745.2 | NP_001307674.1 | |||||
| NM_058187.5 | NP_478067.2 | |||||
| NR_104472.2 | ||||||
| NR_135467.2 | ||||||
| XM_005261019.5 | XP_005261076.1 | |||||
| XM_006724038.4 | XP_006724101.1 | |||||
| XM_011529669.4 | XP_011527971.1 | |||||
| XM_017028418.2 | XP_016883907.1 | |||||
| XM_017028419.2 | XP_016883908.1 | |||||
| XM_017028420.2 | XP_016883909.1 | |||||
| XM_017028422.2 | XP_016883911.1 | |||||
| XM_017028423.2 | XP_016883912.1 | |||||
| XM_047440933.1 | XP_047296889.1 | |||||
| XM_047440934.1 | XP_047296890.1 | |||||
| GenBank mRNA | AF358258.1 | - | ||||
| AI093702.1 | - | |||||
| AI290677.1 | - | |||||
| AK125013.1 | - | |||||
| AK126660.1 | - | |||||
| AL543462.3 | - | |||||
| AW957938.1 | - | |||||
| AY040087.1 | - | |||||
| AY358787.1 | - | |||||
| BC038710.1 | - | |||||
| BM836911.1 | - | |||||
| BU175692.1 | - | |||||
| BX342316.2 | - | |||||
| BX373211.1 | - | |||||
| CA412371.1 | - | |||||