Assay Details
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000594.3 | NP_000585.2 | ||||
GenBank mRNA | AB451492.1 | - | ||||
AF043342.1 | - | |||||
AF098751.1 | - | |||||
AJ227911.1 | - | |||||
AJ251878.1 | - | |||||
AJ251879.1 | - | |||||
AK314960.1 | - | |||||
BC028148.1 | - | 5217636 | ||||
BI908079.1 | - | |||||
BP215875.1 | - | |||||
CA306559.1 | - | |||||
ENST00000449264.0 | - | |||||
FJ795028.1 | - | |||||
HQ201306.2 | - | |||||
M10988.1 | - | |||||
X01394.1 | - |
