Search
Search

BRAF| Gene Symbol: BRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_004333.4 | NP_004324.2 | ||||
| XM_005250045.2 | XP_005250102.1 | |||||
| XM_017012558.1 | XP_016868047.1 | |||||
| XM_017012559.1 | XP_016868048.1 | |||||
| GenBank mRNA | AA252358.1 | - | ||||
| AA834931.1 | - | |||||
| AI203435.1 | - | |||||
| AJ276310.1 | - | |||||
| AK054801.1 | - | |||||
| AK094023.1 | - | |||||
| AK098095.1 | - | |||||
| AK124257.1 | - | |||||
| AK314346.1 | - | |||||
| AW971583.1 | - | |||||
| BC038966.1 | - | |||||
| BC101757.1 | - | 8069263 | ||||
| BC112079.1 | - | 8327547 | ||||
| BM263585.1 | - | |||||
| DB454480.1 | - | |||||
| DN996440.1 | - | |||||
| ENST00000288602.0 | - | |||||
| HM437233.1 | - | |||||
| M21001.1 | - | |||||
| M95712.2 | - | |||||