Search
Search

DPY19L1| Gene Symbol: DPY19L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001366673.1 | NP_001353602.1 | ||||
| NM_015283.2 | NP_056098.1 | |||||
| XM_011515246.4 | XP_011513548.1 | |||||
| GenBank mRNA | AB020684.1 | - | ||||
| AJ011911.1 | - | |||||
| BC029591.1 | - | |||||
| BC050318.1 | - | |||||
| BC064476.1 | - | |||||
| DQ287932.1 | - | |||||