Search
Search

DNM1| Gene Symbol: DNM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001005336.3 | NP_001005336.1 | ||||
| NM_001288737.2 | NP_001275666.1 | |||||
| NM_001288738.2 | NP_001275667.1 | |||||
| NM_001288739.2 | NP_001275668.1 | |||||
| NM_001374269.1 | NP_001361198.1 | |||||
| NM_004408.4 | NP_004399.2 | |||||
| GenBank mRNA | AB209124.1 | - | ||||
| AF035321.1 | - | |||||
| AI660320.1 | - | |||||
| AK096629.1 | - | |||||
| AK127038.1 | - | |||||
| AK225179.1 | - | |||||
| AK295101.1 | - | |||||
| AK296326.1 | - | |||||
| AK308793.1 | - | |||||
| AK316235.1 | - | |||||
| BC050279.1 | - | |||||
| BC063850.1 | - | |||||
| BG396820.1 | - | |||||
| DA221568.1 | - | |||||
| DA510154.1 | - | |||||
| L07807.1 | - | |||||
| L07808.1 | - | |||||
| L07809.1 | - | |||||
| L07810.1 | - | |||||