Assay Details
Gene Symbol: JUN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002228.3 | NP_002219.1 | ||||
GenBank mRNA | BC002646.2 | - | ||||
BC006175.1 | - | 4053956 | ||||
BC009874.2 | - | |||||
BC068522.1 | - | 5267423 | ||||
BG831943.1 | - | |||||
BI793062.1 | - | |||||
BT019759.1 | - | |||||
BU782554.1 | - | |||||
CD370800.1 | - | |||||
CR541724.1 | - | |||||
CR541752.1 | - | |||||
ENST00000371222.0 | - |
