Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TGFA EBGEX Assays ›
          Gene Symbol:
          TGFA
          Gene Name:
          transforming growth factor alpha
          Gene Aliases:
          TFGA
          Chromosome Location:
          Chr.2: 70447284 - 70553826 on Build GRCh38
          UniGene ID:
          Hs.170009
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Type:
          4
          Detection Label:
          Alexa Fluor 488
          ProductType:
          ViewRNA Cell Plus Probe Set
          Assay ID VA4-3083444-VCP
          Size
          Availability Made To Order
          Catalog # VX-01
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TGFA
          Interrogated Sequence Translated Protein IMAGE Clone ID
          RefSeq NM_001099691.3 NP_001093161.1
          NM_001308158.2 NP_001295087.1
          NM_001308159.2 NP_001295088.1
          NM_003236.4 NP_003227.1
          GenBank mRNA AF149096.1 -
          AF149097.1 -
          AF149098.1 -
          AK290151.1 -
          AK309066.1 -
          AK309220.1 -
          AK312899.1 -
          AY548763.1 -
          BC005308.1 -
          BI760767.1 -
          BM453873.1 -
          BT006833.1 -
          CD369689.1 -
          CN274505.1 -
          DA216480.1 -
          DA229819.1 -
          DA373586.1 -
          DA462271.1 -
          K03222.1 -
          M31172.1 -
          X70340.1 -

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          cell surface receptor signaling pathway
          epidermal growth factor receptor signaling pathway
          positive regulation of cell proliferation
          intracellular signal transduction
          ERBB2-EGFR signaling pathway
          positive regulation of MAPK cascade
          positive regulation of mitotic nuclear division
          positive regulation of epithelial cell proliferation
          positive regulation of cell division
          mammary gland alveolus development
          hepatocyte proliferation
          receptor binding
          epidermal growth factor receptor binding
          protein binding
          growth factor activity
          transmembrane receptor protein tyrosine kinase activator activity
          receptor agonist activity

          Back To Top
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline