Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MERTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_006343.2 | NP_006334.2 | ||||
XM_005263565.4 | XP_005263622.1 | |||||
XM_005263568.4 | XP_005263625.1 | |||||
XM_011510490.2 | XP_011508792.1 | |||||
XM_017003164.1 | XP_016858653.1 | |||||
XM_017003165.1 | XP_016858654.1 | |||||
GenBank mRNA | AK026681.1 | - | ||||
AK294196.1 | - | |||||
AK308695.1 | - | |||||
AK316584.1 | - | |||||
AL705756.1 | - | |||||
AW627665.1 | - | |||||
BC114917.1 | - | 40068588 | ||||
BC114918.1 | - | 40012335 | ||||
BM719997.1 | - | |||||
BX474860.1 | - | |||||
CA454601.1 | - | |||||
CN371299.1 | - | |||||
CN371300.1 | - | |||||
D16316.1 | - | |||||
U08023.1 | - |