Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other XPA EBGEX Assays ›
          Gene Symbol:
          XPA
          Gene Name:
          XPA, DNA damage recognition and repair factor
          Gene Aliases:
          XP1, XPAC
          Chromosome Location:
          Chr.9: 97654398 - 97697340 on Build GRCh38
          UniGene ID:
          Hs.654364
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Type:
          4
          Detection Label:
          Alexa Fluor 488
          ProductType:
          ViewRNA Cell Plus Probe Set
          Assay ID VA4-3088757-VCP
          Size
          Availability Made To Order
          Catalog # VX-01
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: XPA
          Interrogated Sequence Translated Protein IMAGE Clone ID
          RefSeq NM_000380.4 NP_000371.1
          NM_001354975.2 NP_001341904.1
          NR_027302.2
          NR_149091.2
          NR_149092.2
          NR_149093.2
          NR_149094.2
          XM_006717278.2 XP_006717341.1
          GenBank mRNA AI961077.1 -
          AL531629.3 -
          BC014965.1 -
          BT019518.1 -
          CB155090.1 -
          D14533.1 -
          KC899693.1 -
          S89853.1 -

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          nucleotide-excision repair, DNA damage recognition
          DNA repair
          base-excision repair
          nucleotide-excision repair
          cellular response to DNA damage stimulus
          response to oxidative stress
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to UV
          response to toxic substance
          UV protection
          regulation of autophagy
          protein localization to nucleus
          multicellular organism growth
          positive regulation of transcription initiation from RNA polymerase II promoter
          UV-damage excision repair
          nucleotide-excision repair involved in interstrand cross-link repair
          DNA binding
          damaged DNA binding
          protein binding
          zinc ion binding
          protein domain specific binding
          protein homodimerization activity
          metal ion binding
          sequence-specific double-stranded DNA binding

          Back To Top
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-v4tl5:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline