Search
Search

NGFR| Gene Symbol: NGFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_002507.3 | NP_002498.1 | ||||
| GenBank mRNA | AK303278.1 | - | ||||
| AK313654.1 | - | |||||
| BC050309.1 | - | 5263715 | ||||
| BC052797.1 | - | |||||
| BQ639556.1 | - | |||||
| CK299927.1 | - | |||||
| DA115173.1 | - | |||||
| ENST00000172229.0 | - | |||||
| M14764.1 | - | |||||