Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_003954.4 | NP_003945.2 | ||||
XM_011525441.2 | XP_011523743.1 | |||||
GenBank mRNA | AA564389.1 | - | ||||
AJ008135.1 | - | |||||
AJ008136.1 | - | |||||
AJ008137.1 | - | |||||
AJ008138.1 | - | |||||
AJ008139.1 | - | |||||
AJ008140.1 | - | |||||
AJ008141.1 | - | |||||
AJ008142.1 | - | |||||
AJ008143.1 | - | |||||
AJ008144.1 | - | |||||
AJ008145.1 | - | |||||
AJ008146.1 | - | |||||
AJ008147.1 | - | |||||
AJ008148.1 | - | |||||
AJ008149.1 | - | |||||
AJ008150.1 | - | |||||
AJ008151.1 | - | |||||
AJ008152.1 | - | |||||
AJ008153.1 | - | |||||
AJ008156.1 | - | |||||
AJ008157.1 | - | |||||
AJ008158.1 | - | |||||
AJ008159.1 | - | |||||
AJ008160.1 | - | |||||
AJ008161.1 | - | |||||
AJ008162.1 | - | |||||
AJ008163.1 | - | |||||
AK123696.1 | - | |||||
AK131438.1 | - | |||||
AK290203.1 | - | |||||
AK313085.1 | - | |||||
BC022296.1 | - | |||||
BC035576.1 | - | 5497185 | ||||
BE645094.1 | - | |||||
BU623799.1 | - | |||||
CR749592.1 | - | |||||
DA856801.1 | - | |||||
HY096069.1 | - | |||||
HY287427.1 | - | |||||
Y10256.1 | - |