Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001014831.2 | NP_001014831.1 | ||||
NM_001014832.1 | NP_001014832.1 | |||||
NM_001014834.2 | NP_001014834.1 | |||||
NM_001014835.1 | NP_001014835.1 | |||||
NM_005884.3 | NP_005875.1 | |||||
XM_011526316.1 | XP_011524618.1 | |||||
XM_011526317.2 | XP_011524619.1 | |||||
XM_011526318.2 | XP_011524620.1 | |||||
XM_011526319.2 | XP_011524621.1 | |||||
XM_011526320.1 | XP_011524622.1 | |||||
GenBank mRNA | AB032968.1 | - | ||||
AF005046.1 | - | |||||
AJ011855.1 | - | |||||
AK074728.1 | - | |||||
AK291839.1 | - | |||||
AK293517.1 | - | |||||
AK294586.1 | - | |||||
AK300632.1 | - | |||||
AK304651.1 | - | |||||
AL834236.1 | - | |||||
AX775863.1 | - | |||||
AX775865.1 | - | |||||
AX775867.1 | - | |||||
AX775869.1 | - | |||||
AX775871.1 | - | |||||
BC002921.2 | - | 3956856 | ||||
BC011368.1 | - | 3867403 | ||||
BC025282.1 | - | 5421365 | ||||
BC034511.1 | - | 3882622 | ||||
BG773480.1 | - | |||||
BM129363.1 | - | |||||
ENST00000358301.0 | - |