Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001198777.1 | NP_001185706.1 | ||||
NM_001198778.1 | NP_001185707.1 | |||||
NM_001198779.1 | NP_001185708.1 | |||||
NM_001324375.1 | NP_001311304.1 | |||||
NM_001324376.1 | NP_001311305.1 | |||||
NM_003591.3 | NP_003582.2 | |||||
XM_011519743.1 | XP_011518045.1 | |||||
XM_011519744.1 | XP_011518046.1 | |||||
XM_011519745.1 | XP_011518047.1 | |||||
XM_011519747.1 | XP_011518049.1 | |||||
GenBank mRNA | AF126404.1 | - | ||||
AI089201.1 | - | |||||
AK095217.1 | - | |||||
AK294080.1 | - | |||||
AK300491.1 | - | |||||
AK310225.1 | - | |||||
AL832733.1 | - | |||||
BC009591.1 | - | 4104375 | ||||
BC095405.1 | - | |||||
BC110901.1 | - | 6161736 | ||||
DC401811.1 | - | |||||
DC401936.1 | - | |||||
ENST00000374751.0 | - | |||||
U58088.1 | - | |||||
U83410.1 | - |