Search
Search

Fosb| Gene Symbol: Fosb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_008036.2 | NP_032062.1 | ||||
| GenBank mRNA | AI846927.1 | - | ||||
| AK154373.1 | - | |||||
| BC132064.1 | - | 40130341 | ||||
| BC137994.1 | - | 8861013 | ||||
| BC145082.1 | - | |||||
| BU698478.1 | - | |||||
| BY337108.1 | - | |||||
| BY547195.1 | - | |||||
| CF356426.1 | - | |||||
| CK795975.1 | - | |||||