Assay Details
Gene Symbol: Jun | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_010591.2 | NP_034721.1 | ||||
GenBank mRNA | AK132019.1 | - | ||||
AK133722.1 | - | |||||
AK133867.1 | - | |||||
AK135023.1 | - | |||||
AK140911.1 | - | |||||
AK159196.1 | - | |||||
AK159220.1 | - | |||||
AY423853.1 | - | |||||
BC002081.1 | - | 3493248 | ||||
BC021888.1 | - | 3156091 | ||||
BC032086.1 | - | |||||
BC055887.1 | - | |||||
BC094032.1 | - | 4936392 | ||||
BY106462.1 | - | |||||
J04115.1 | - | |||||
X12740.1 | - | |||||
X12761.1 | - |
