Search
Search

Rpl13a| Gene Symbol: Rpl13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_009438.5 | NP_033464.2 | ||||
| GenBank mRNA | AK007609.1 | - | ||||
| AK077512.1 | - | |||||
| AK146318.1 | - | |||||
| AK156035.1 | - | |||||
| AK159882.1 | - | |||||
| AK168784.1 | - | |||||
| AK168903.1 | - | |||||
| AK170030.1 | - | |||||
| AK189288.1 | - | |||||
| AK213140.1 | - | |||||
| AV445288.1 | - | |||||
| BC034450.1 | - | |||||
| BC042392.1 | - | |||||
| BC065799.1 | - | |||||
| BC082289.1 | - | 6486177 | ||||
| BC086896.1 | - | 6776320 | ||||
| CT010169.1 | - | |||||
Set Membership: |
|