Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ENO1 EBGEX Assays ›
Gene Symbol:
Eno1
Gene Name:
enolase 1, alpha non-neuron
Gene Aliases:
0610008I15, AL022784, Eno-1, MBP-1
Chromosome Location:
Chr.4: 150236726 - 150248873 on Build GRCm38
UniGene ID:
Mm.372357
Species:
Mouse
Species Specific ID (Flybase ID):
-
Type:
1
Detection Label:
Alexa Fluor 546
ProductType:
ViewRNA Cell Probe Set
Assay ID
VB1-19720-VC
Size
S: 120 uL
M: 440 uL
L: 1.44 mL
Availability
Made To Order
Catalog #
VX-06
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Eno1
Interrogated Sequence
Translated Protein
IMAGE Clone ID
RefSeq
NM_023119.2
NP_075608.2
GenBank mRNA
AK002322.1
-
AK002336.1
-
AK014335.1
-
AK088788.1
-
AK147987.1
-
AK150161.1
-
AK150192.1
-
AK153321.1
-
AK177990.1
-
AK179805.1
-
AK180361.1
-
AK180697.1
-
AK186626.1
-
AK190122.1
-
AK190135.1
-
AK190320.1
-
AK190571.1
-
AK190754.1
-
AK190836.1
-
AK190994.1
-
AK190997.1
-
AK191162.1
-
AK191266.1
-
AK191332.1
-
AK191599.1
-
AK192110.1
-
AK192771.1
-
AK192774.1
-
AK192856.1
-
AK193395.1
-
AK193470.1
-
AK193472.1
-
AK194042.1
-
AK194050.1
-
AK194112.1
-
AK194157.1
-
AK194259.1
-
AK194312.1
-
AK194383.1
-
AK194493.1
-
AK194536.1
-
AK194989.1
-
AK195184.1
-
AK195411.1
-
AK195458.1
-
AK195645.1
-
AK196007.1
-
AK196033.1
-
AK196278.1
-
AK196762.1
-
AK196851.1
-
AK196981.1
-
AK197056.1
-
AK197091.1
-
AK197358.1
-
AK197441.1
-
AK197783.1
-
AK198034.1
-
AK198256.1
-
AK198771.1
-
AK198922.1
-
AK198960.1
-
AK199002.1
-
AK199229.1
-
AK199273.1
-
AK199319.1
-
AK199394.1
-
AK199498.1
-
AK199950.1
-
AK199973.1
-
AK200096.1
-
AK201180.1
-
AK201244.1
-
AK201655.1
-
AK202010.1
-
AK202164.1
-
AK202376.1
-
AK202428.1
-
AK202456.1
-
AK202490.1
-
AK202509.1
-
AK202755.1
-
AK202890.1
-
AK202928.1
-
AK203109.1
-
AK203149.1
-
AK203172.1
-
AK203403.1
-
AK203497.1
-
AK203510.1
-
AK203592.1
-
AK203740.1
-
AK204087.1
-
AK204111.1
-
AK204315.1
-
AK204457.1
-
AK204692.1
-
AK204945.1
-
AK204949.1
-
AK206473.1
-
AK207175.1
-
AK207191.1
-
AK207837.1
-
AK208860.1
-
AK209576.1
-
AK211315.1
-
AK211960.1
-
AK214077.1
-
AK217406.1
-
AK218216.1
-
AK219016.1
-
AK219721.1
-
BC003891.1
-
3592683
BC003934.1
-
BC010685.1
-
3599051
BC024644.1
-
3583123
BC039179.1
-
4501041
BC056611.1
-
4948983
BC083334.1
-
6414729
BC085098.1
-
5376359
BU526382.1
-
X52379.1
-
Back To Top
More Information
Panther Classification:
Molecular Function -
lyase
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of binding
cellular response to acid chemical
RNA binding
phosphopyruvate hydratase activity
protein homodimerization activity
poly(A) RNA binding
protein heterodimerization activity
GTPase binding
Back To Top
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline