Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_010721.2 | NP_034851.2 | ||||
GenBank mRNA | AK079511.1 | - | ||||
AK089110.1 | - | |||||
AK132252.1 | - | |||||
AK180884.1 | - | |||||
BC052729.1 | - | 6816118 | ||||
BC058392.1 | - | 6816118 | ||||
M35153.1 | - | |||||
X16705.1 | - |