Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001165902.1 | NP_001159374.1 | ||||
NM_007614.3 | NP_031640.1 | |||||
GenBank mRNA | AK018515.1 | - | ||||
AK020013.1 | - | |||||
AK035311.1 | - | |||||
AK132398.1 | - | |||||
AK133656.1 | - | |||||
AK135743.1 | - | |||||
AK178155.1 | - | |||||
AK179148.1 | - | |||||
AK179260.1 | - | |||||
AK180997.1 | - | |||||
AK184861.1 | - | |||||
AK189807.1 | - | |||||
AK190684.1 | - | |||||
AK191852.1 | - | |||||
AK196236.1 | - | |||||
AK196547.1 | - | |||||
AK197513.1 | - | |||||
AK197664.1 | - | |||||
AK197753.1 | - | |||||
AK199952.1 | - | |||||
AK201646.1 | - | |||||
AK201809.1 | - | |||||
AK202371.1 | - | |||||
AK203436.1 | - | |||||
AK203824.1 | - | |||||
AK204053.1 | - | |||||
AK208830.1 | - | |||||
AK209047.1 | - | |||||
AK209437.1 | - | |||||
AK209908.1 | - | |||||
AK210588.1 | - | |||||
AK211747.1 | - | |||||
AK214825.1 | - | |||||
AK219273.1 | - | |||||
AK219949.1 | - | |||||
AY751549.1 | - | |||||
BC006739.1 | - | |||||
BC043078.1 | - | |||||
BC043481.1 | - | |||||
BC048153.1 | - | 5709247 | ||||
BC053065.1 | - | 5709247 | ||||
BY766878.1 | - | |||||
M90364.1 | - |