Assay Details
Gene Symbol: Nfkbia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_010907.2 | NP_035037.2 | ||||
GenBank mRNA | AI462015.1 | - | ||||
AK089215.1 | - | |||||
AK151115.1 | - | |||||
AK151608.1 | - | |||||
AK161622.1 | - | |||||
BC046754.1 | - | 5693175 | ||||
CF379853.1 | - | |||||
U36277.1 | - |
