Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MFGE8 EBGEX Assays ›
          Gene Symbol:
          Mfge8
          Gene Name:
          milk fat globule-EGF factor 8 protein
          Gene Aliases:
          AA408458, AI325141, MFG-E8, MP47, Mfgm, P47, SED1, lactadherin
          Chromosome Location:
          Chr.7: 79133768 - 79149060 on Build GRCm38
          UniGene ID:
          Mm.1451
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Type:
          10
          Detection Label:
          Alexa Fluor 750
          ProductType:
          ViewRNA Cell Probe Set
          Assay ID VB10-3283456-VC
          Size
          Availability Made To Order
          Catalog # VX-01
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mfge8
          Interrogated Sequence Translated Protein IMAGE Clone ID
          RefSeq NM_001045489.1 NP_001038954.1
          NM_008594.2 NP_032620.2
          GenBank mRNA AB021130.2 -
          AB025280.1 -
          AK087555.1 -
          AK089211.1 -
          AK145183.1 -
          AK148371.1 -
          AK152088.1 -
          AK155047.1 -
          AK169997.1 -
          AK170935.1 -
          AK171143.1 -
          AK178722.1 -
          AK179489.1 -
          AK181224.1 -
          AK182601.1 -
          AK185944.1 -
          AK189618.1 -
          AK189622.1 -
          AK190455.1 -
          AK190819.1 -
          AK192581.1 -
          AK193016.1 -
          AK193609.1 -
          AK193753.1 -
          AK193868.1 -
          AK194123.1 -
          AK194145.1 -
          AK194326.1 -
          AK194803.1 -
          AK195127.1 -
          AK195377.1 -
          AK195556.1 -
          AK195675.1 -
          AK196442.1 -
          AK196948.1 -
          AK197795.1 -
          AK199381.1 -
          AK200279.1 -
          AK200283.1 -
          AK200528.1 -
          AK200809.1 -
          AK200926.1 -
          AK201191.1 -
          AK202028.1 -
          AK202981.1 -
          AK203389.1 -
          AK204233.1 -
          AK204253.1 -
          AK204278.1 -
          AK205061.1 -
          AK205167.1 -
          AK206323.1 -
          AK206818.1 -
          AK206986.1 -
          AK207561.1 -
          AK207636.1 -
          AK209209.1 -
          AK210333.1 -
          AK210816.1 -
          AK212128.1 -
          AK212659.1 -
          AK213407.1 -
          AK214404.1 -
          AK218369.1 -
          AK219560.1 -
          AK219564.1 -
          AK219710.1 -
          BC003892.1 - 3593310
          BC003904.1 - 3601565
          BC018577.1 -
          BC037475.1 -
          CD776821.1 -
          M38337.1 -
          Y11684.2 -

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          phagocytosis, recognition
          phagocytosis, engulfment
          cell adhesion
          single fertilization
          positive regulation of phagocytosis
          positive regulation of apoptotic cell clearance
          phosphatidylserine binding
          integrin binding
          phosphatidylethanolamine binding

          Back To Top
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline