Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crebbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001025432.1 | NP_001020603.1 | ||||
GenBank mRNA | AK048818.1 | - | ||||
AK051071.1 | - | |||||
AK136057.1 | - | |||||
AK162593.1 | - | |||||
BC010297.1 | - | |||||
BC072594.1 | - | |||||
BU705617.1 | - | |||||
S66385.1 | - |