Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_013518.4 | NP_038546.2 | ||||
GenBank mRNA | AI414273.1 | - | ||||
AK041815.1 | - | |||||
AK079617.1 | - | |||||
AK158584.1 | - | |||||
AK158782.1 | - | |||||
AK164411.1 | - | |||||
BC099872.1 | - | 40040091 | ||||
BC099873.1 | - | 40040090 | ||||
BC099874.1 | - | 40040089 | ||||
BC099875.1 | - | 40040088 | ||||
BC125237.1 | - | 40040117 | ||||
BX512213.1 | - | |||||
D38258.1 | - | |||||
HM988990.1 | - | |||||
S82023.1 | - | |||||
U33535.1 | - |