Search
Search

Chst15| Gene Symbol: Chst15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_029935.5 | NP_084211.2 | ||||
| GenBank mRNA | AB187269.1 | - | ||||
| AF320786.1 | - | |||||
| AK019474.1 | - | |||||
| AK031745.1 | - | |||||
| AK031995.1 | - | |||||
| AK035637.1 | - | |||||
| AK043803.1 | - | |||||
| AK082318.1 | - | |||||
| AK083614.1 | - | |||||
| AK122324.1 | - | |||||
| BC031443.1 | - | 4240286 | ||||