Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mapk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_016700.4 | NP_057909.1 | ||||
GenBank mRNA | AB005663.1 | - | ||||
AB206575.1 | - | |||||
AB219977.1 | - | |||||
AI849689.1 | - | |||||
AK030767.1 | - | |||||
AK032486.1 | - | |||||
AK039129.1 | - | |||||
AK039254.1 | - | |||||
AK043102.1 | - | |||||
AK044768.1 | - | |||||
AK047936.1 | - | |||||
AK053062.1 | - | |||||
AK085745.1 | - | |||||
AK139498.1 | - | |||||
AK162915.1 | - | |||||
BB619635.1 | - | |||||
BC053027.1 | - | 5694771 |