Search
Search

Irf2| Gene Symbol: Irf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_008391.4 | NP_032417.3 | ||||
| GenBank mRNA | AK036652.1 | - | ||||
| AK087926.1 | - | |||||
| AK132099.1 | - | |||||
| AK155025.1 | - | |||||
| AK158642.1 | - | |||||
| BC006577.1 | - | 3586586 | ||||
| BC110666.1 | - | 4918447 | ||||
| J03168.1 | - | |||||