Search
Search

Baz2b| Gene Symbol: Baz2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001001182.3 | NP_001001182.2 | ||||
| GenBank mRNA | AK029231.1 | - | ||||
| AK029386.1 | - | |||||
| AK030867.1 | - | |||||
| AK047236.1 | - | |||||
| AK048335.1 | - | |||||
| AK079809.1 | - | |||||
| AK083663.1 | - | |||||
| AK134250.1 | - | |||||
| AK137989.1 | - | |||||
| AK143439.1 | - | |||||
| AK147737.1 | - | |||||
| AK166079.1 | - | |||||
| AK171175.1 | - | |||||
| AK173180.1 | - | |||||
| BC027336.1 | - | |||||
| BC042646.1 | - | |||||
| BC053917.1 | - | |||||
| BC150814.1 | - | 9087725 | ||||
| BY740822.1 | - | |||||
| CA319831.1 | - | |||||
| CB247705.1 | - | |||||
| CN534605.1 | - | |||||