Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001285453.1 | NP_001272382.1 | ||||
NM_007522.3 | NP_031548.1 | |||||
GenBank mRNA | AK029400.1 | - | ||||
AK131666.1 | - | |||||
AK151792.1 | - | |||||
AK169002.1 | - | |||||
BC006762.1 | - | 3257589 | ||||
L37296.1 | - |