Search
Search

Ctu2| Gene Symbol: Ctu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_153775.2 | NP_722470.2 | ||||
| GenBank mRNA | AK012440.1 | - | ||||
| AK090349.1 | - | |||||
| AK090391.1 | - | |||||
| AK154995.1 | - | |||||
| AK172353.1 | - | |||||
| AK188865.1 | - | |||||
| AK206078.1 | - | |||||
| AK211906.1 | - | |||||
| AK215943.1 | - | |||||
| BC036332.1 | - | 3156706 | ||||
| CJ067802.1 | - | |||||