Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_029402.3 | NP_083678.2 | ||||
GenBank mRNA | AK004875.2 | - | ||||
AK016520.1 | - | |||||
AK028589.1 | - | |||||
AK144693.1 | - | |||||
AK160597.1 | - | |||||
AK162910.1 | - | |||||
AK204549.1 | - | |||||
AK220287.1 | - | |||||
BC025902.1 | - | |||||
BC026779.1 | - | 4018298 | ||||
BC027428.1 | - | 3983461 |