Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map4k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_133407.1 | NP_596898.1 | ||||
XM_006239656.2 | XP_006239718.1 | |||||
GenBank mRNA | AA819812.1 | - | ||||
AF312224.1 | - | |||||
AI502900.1 | - | |||||
CK474868.1 | - | |||||
DV716943.1 | - |