Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MKK7 EBGEX Assays ›
Gene Symbol:
MKK7
Gene Name:
MAP kinase kinase 7
Gene Aliases:
AT1G18350, ATMKK7, BUD1, BUSHY AND DWARF 1, F15H18.14, F15H18_14, MAP KINASE KINASE7, MAP kinase kinase 7
Chromosome Location:
Chr.1: 6315636 - 6316738 on Build TAIR10.1
UniGene ID:
At.28338
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Type:
1
Detection Label:
Alexa Fluor 546
ProductType:
ViewRNA Cell Probe Set
Assay ID
VF1-4016213-VC
Size
M: 440 uL
L: 1.44 mL
Availability
Made To Order
Catalog #
VX-01
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: MKK7
Interrogated Sequence
Translated Protein
IMAGE Clone ID
RefSeq
NM_101693.3
NP_173271.1
GenBank mRNA
DQ185389.1
-
DQ446261.1
-
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
defense response to oomycetes
systemic acquired resistance, salicylic acid mediated signaling pathway
auxin polar transport
defense response to bacterium
MAP kinase kinase activity
kinase activity
Back To Top
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-z8nhc:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0