Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SHG EBGEX Assays ›
Gene Symbol:
shg
Gene Name:
shotgun
Gene Aliases:
CADH, CG3722, CT12481, Cad, CadE, Cadh, D E-cad, D-CAD2, D-cad, DCAD, DCAD2, DCad, DCad2, DE, DE Cad, DE Cadh, DE-CAD, DE-CAD2, DE-Cad, DE-Cad2, DE-Cadh, DE-Cadherin, DE-cad, DE-cadh, DE-cadherin, DEC, DECAD, DECad, DECadh, DE[cyto], DEcad, De-Cad, Dmel\CG3722, Dmel_CG3722, E-CAD, E-Ca, E-Cad, E-Cadherin, E-cad, E-cadherin, ECAD, ECad, ECad2, ECadh, Ecad, N-cad, Shg, cad, cadh, com5, dEcad, de-cad, e-cad, ecad, gp150, l(2)10469, l(2)k03401, l(2)k10220, shg/DE-Cad
Chromosome Location:
Chr.2R: 21049654 - 21057197 on Build Release_6_plus_ISO1_MT
UniGene ID:
-
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0003391
Type:
1
Detection Label:
Alexa Fluor 546
ProductType:
ViewRNA Cell Plus Probe Set
Assay ID
VF1-4149538-VCP
Size
M: 440 uL
L: 1.44 mL
Availability
Made To Order
Catalog #
VX-01
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: shg
Interrogated Sequence
Translated Protein
IMAGE Clone ID
RefSeq
NM_057374.3
NP_476722.1
Back To Top
More Information
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
morphogenesis of a polarized epithelium
compound eye morphogenesis
optic lobe placode development
outflow tract morphogenesis
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
germ cell development
border follicle cell migration
ventral furrow formation
segment specification
nervous system development
axonogenesis
brain development
open tracheal system development
salivary gland morphogenesis
gonadal mesoderm development
heart development
head involution
germ cell migration
asymmetric cell division
gonad development
gastrulation involving germ band extension
ommatidial rotation
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
cell projection assembly
oocyte localization involved in germarium-derived egg chamber formation
somatic stem cell population maintenance
hemocyte migration
branch fusion, open tracheal system
maintenance of epithelial integrity, open tracheal system
cell competition in a multicellular organism
gonad morphogenesis
female germ-line stem cell population maintenance
wound healing
cell-cell adhesion mediated by cadherin
apical protein localization
zonula adherens assembly
oogenesis
defense response to fungus
determination of digestive tract left/right asymmetry
cell elongation involved in imaginal disc-derived wing morphogenesis
cell-cell adhesion
male germline stem cell symmetric division
positive regulation of border follicle cell migration
calcium ion binding
protein binding
myosin binding
protein homodimerization activity
Back To Top
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline