Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PP1-87B EBGEX Assays ›
Gene Symbol:
Pp1-87B
Gene Name:
Protein phosphatase 1 at 87B
Gene Aliases:
87B, CG5650, DmPp1-87B, Dmel\CG5650, Dmel_CG5650, PP-1alpha, PP1, PP1 87B, PP1 beta87b, PP1-87B, PP187B, PP1A87B, PP1B, PP1a, PP1alpha-87B, PP1alpha87B, PP1alpha[[2]], PP1c, PP1c 87B, PPI 87B, Pp-1alpha, Pp1, Pp1-87Bb, Pp1alpha-87B, Ppl-87B, Su(Var)3-6, Su(var)(3)6, Su(var)3-6, Su-var(3)6, Suvar(3)6, ck19, dPP1beta, l(3)87Bg, l(3)ck19, l(3)j6E7, pp1-87, pp1-87B, pp187B, su-var(3)6
Chromosome Location:
Chr.3R: 12423621 - 12425917 on Build Release_6_plus_ISO1_MT
UniGene ID:
Dm.1623
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0004103
Type:
1
Detection Label:
FastRed
ProductType:
ViewRNA Tissue Probe Set
Assay ID
VF1-4151511-VT
Size
M: 440 uL
L: 1.44 mL
Availability
Made To Order
Catalog #
VX-01
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Pp1-87B
Interrogated Sequence
Translated Protein
IMAGE Clone ID
RefSeq
NM_080198.3
NP_524937.1
GenBank mRNA
AY061063.1
-
X15583.1
-
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cell cycle
protein dephosphorylation
chromosome segregation
mitotic metaphase plate congression
nervous system development
axon guidance
learning or memory
adult locomotory behavior
olfactory learning
visual learning
chromosome condensation
locomotion
positive regulation of Ras protein signal transduction
oogenesis
spindle assembly
positive regulation of canonical Wnt signaling pathway
beta-catenin destruction complex disassembly
protein serine/threonine phosphatase activity
protein binding
myosin phosphatase activity
Back To Top
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline