Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FZ EBGEX Assays ›
Gene Symbol:
fz
Gene Name:
frizzled
Gene Aliases:
CG17697, CG3646, DFZ1, DFz, DFz1, Dfz, Dfz1, Dm Fz1, Dmel\CG17697, Dmel_CG17697, FZ, FZ1, Frizzled, Frz, Fz, Fz1, dFz, frz, fz1, fz[[1]]
Chromosome Location:
Chr.3L: 14274343 - 14368639 on Build Release_6_plus_ISO1_MT
UniGene ID:
Dm.1456
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0001085
Type:
4
Detection Label:
Alexa Fluor 488
ProductType:
ViewRNA Cell Probe Set
Assay ID
VF4-4151341-VC
Size
M: 440 uL
L: 1.44 mL
Availability
Made To Order
Catalog #
VX-01
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: fz
Interrogated Sequence
Translated Protein
IMAGE Clone ID
RefSeq
NM_001274907.1
NP_001261836.1
NM_080073.3
NP_524812.1
GenBank mRNA
AY051808.1
-
X54646.1
-
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
establishment of planar polarity
establishment of imaginal disc-derived wing hair orientation
morphogenesis of a polarized epithelium
compound eye morphogenesis
homophilic cell adhesion via plasma membrane adhesion molecules
establishment or maintenance of cell polarity
G-protein coupled receptor signaling pathway
border follicle cell migration
segment polarity determination
salivary gland morphogenesis
R3/R4 cell fate commitment
midgut development
asymmetric cell division
imaginal disc-derived wing margin morphogenesis
ommatidial rotation
sensory organ precursor cell fate determination
establishment of cell polarity
regulation of actin filament bundle assembly
regulation of tube length, open tracheal system
regulation of hemocyte proliferation
chitin-based larval cuticle pattern formation
imaginal disc-derived wing hair organization
imaginal disc-derived wing hair site selection
non-canonical Wnt signaling pathway
establishment of ommatidial planar polarity
cardioblast cell fate specification
positive regulation of axon extension
R3/R4 cell differentiation
neuron fate specification
axon extension
canonical Wnt signaling pathway
germ-line stem-cell niche homeostasis
positive regulation of axon guidance
Wnt-protein binding
Wnt-activated receptor activity
Back To Top
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline