Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001103936.2 | NP_001097406.1 | ||||
NM_001169775.2 | NP_001163246.1 | |||||
NM_001169776.2 | NP_001163247.1 | |||||
NM_001169777.2 | NP_001163248.1 | |||||
NM_001169778.2 | NP_001163249.1 | |||||
NM_001299775.1 | NP_001286704.1 | |||||
NM_137796.3 | NP_611640.3 | |||||
NM_166507.2 | NP_726159.1 | |||||
NM_166508.2 | NP_726160.1 | |||||
NM_166509.2 | NP_726161.1 | |||||
NM_166510.2 | NP_726162.1 | |||||
NM_166511.2 | NP_726163.1 | |||||
GenBank mRNA | AY094731.1 | - |