Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101725754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | XM_004843076.3 | XP_004843133.1 | ||||
XM_004843077.3 | XP_004843134.1 | |||||
XM_013071580.2 | XP_012927034.1 |