Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084073.1 | NP_001077542.1 | 1 - 2 | 386 | 127 | |
NM_101437.5 | NP_173021.1 | 1 - 2 | 450 | 127 | ||
GenBank mRNA | BT002481.1 | - | 1 - 2 | 386 | 127 | |
M81892.1 | - | 1 - 2 | 416 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |