Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp70b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101471.3 | NP_173055.1 | 1 | 2249 | 119 | |
GenBank mRNA | BT001988.1 | - | 1 | 2250 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |