Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332209.1 | NP_001319017.1 | 5 - 6 | 1056 | 70 | |
NM_101475.3 | NP_173059.1 | 5 - 6 | 1068 | 70 | ||
GenBank mRNA | AF487269.1 | - | 5 - 6 | 867 | 70 | |
BT025624.1 | - | 5 - 6 | 867 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |