Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATKDSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035978.2 | NP_001031055.1 | 13 - 14 | 929 | 70 | |
NM_001084078.1 | NP_001077547.1 | 13 - 14 | 869 | 70 | ||
NM_001198082.1 | NP_001185011.1 | 12 - 13 | 1005 | 70 | ||
NM_101500.4 | NP_173084.1 | 12 - 13 | 1118 | 70 | ||
GenBank mRNA | AK175407.1 | - | 13 - 14 | 901 | 70 | |
BT010438.1 | - | 12 - 13 | 826 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |