Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198089.1 | NP_001185018.1 | 6 - 7 | 1166 | 100 | |
NM_001332274.1 | NP_001321275.1 | 5 - 6 | 1213 | 100 | ||
NM_101564.4 | NP_173147.2 | 5 - 6 | 1213 | 100 | ||
GenBank mRNA | BT046180.1 | - | 5 - 6 | 875 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |