Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101619.2 | NP_173200.1 | 2 - 3 | 248 | 81 | |
GenBank mRNA | BT004292.1 | - | 2 - 3 | 219 | 81 | |
BT005522.1 | - | 2 - 3 | 124 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |