Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101647.5 | NP_173226.2 | 3 - 4 | 1309 | 79 | |
GenBank mRNA | AY080792.1 | - | 3 - 4 | 1088 | 79 | |
AY150457.1 | - | 3 - 4 | 841 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |