Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101647.5 | NP_173226.2 | 7 - 8 | 1849 | 68 | |
GenBank mRNA | AY080792.1 | - | 7 - 8 | 1628 | 68 | |
AY150457.1 | - | 7 - 8 | 1381 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |