Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101716.7 | NP_564057.4 | 1 - 2 | 532 | 97 | |
GenBank mRNA | AY050967.1 | - | 1 - 2 | 373 | 97 | |
AY062444.1 | - | 1 - 2 | 437 | 97 | ||
KY906027.1 | - | 1 - 2 | 157 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |